Learn More
Selleck Chemical LLC 8-OH-DPAT
SDP
Supplier:  Selleck Chemical LLC S8447100MG
Encompass_Preferred
8-OH-DPAT (8-Hydroxy-DPAT) is a kind of classic 5-HT1A agonist with the pIC50 of 8 19 It has a selectivity of almost-1000 fold for a subtype of the 5-HT1 binding site Its biological half-life is 1 5 hous
Catalog No. 50-000-02205