Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-Quinolinecarboxylic acid | 86-59-9 | 173.17 | 10 G
SDP

Supplier:  Medchemexpress LLC HYW00737110G

Encompass_Preferred

Quinoline-8-carboxylic acid is a chemical compound primarily known for its herbicidal properties. This product is intended for research use only and is not for patient administration.

  • Appears as a solid
  • Color ranges from light yellow to brown
  • High purity of 99.46%
  • Molecular weight of 173.17
  • Molecular formula C10H7NO2
  • Soluble in DMSO
  • Powder can be stored at -20°C for 3 years or 4°C for 2 years
  • Solution can be stored at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-60360


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.