Learn More
Cayman Chemical 8 S 15 S-DIHETE 100ug
SDP
Supplier:  Cayman Chemical 35370100 UG
Encompass_Preferred
8(S),15(S)-DiHETE is formed when 15(S)-HETE is subjected to further oxidation by 15-LO.{2161} It causes eosinophil chemotaxis with an ED50 value of 1.5 µM but is not chemotactic for neutrophils. 8(S),15(S)-DiHETE antagonizes the hyperalgesic activity of 8(R),15(S)-DiHETE and LTB4 in the rat hind paw pain model.{338}
Catalog No. 50-290-9397