Learn More
Medchemexpress LLC 8-Shogaol | 36700-45-5 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN24351MG
Encompass_Preferred
8-Shogaol is a stimulating compound found in ginger that exhibits antiplatelet, anti-cancer, and anti-inflammatory activities. It inhibits COX-2, leading to a decline in human leukemia cells. Additionally, it selectively targets TAK1, suppressing the IKK, Akt, and MAPK signaling pathways, and can reverse synovitis synovial in air dampness (RA).
  • Exhibits antiplatelet, anti-cancer, and anti-inflammatory activities
  • Inhibits COX-2 leading to a decline in human leukemia cells
  • Selectively targets TAK1 suppressing IKK, Akt, and MAPK signal pathways
  • Reverses synovitis in rheumatoid arthritis (RA)
  • Shows cytotoxic effects on HL-60 cell growth in a concentration-dependent manner
  • Reduces paw thickness and improves walking performance in a rat model of adjuvant-induced arthritis (AIA)
Catalog No. 50-004-21302