Learn More
Medchemexpress LLC 8-shogaol | 36700-45-5 | MFCD21333718 | 99.9% | 304.42 g/mol | C19H28O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN243510MG
Encompass_Preferred
8-Shogaol is a ginger-derived bioactive phenolic compound reported to inhibit cyclooxygenase enzymes and to exhibit antiplatelet, anti-inflammatory, and anticancer activities. It is supplied as a high-purity analytical standard for research use and is accompanied by documentation to support assay development and safety compliance.
  • Derived from ginger and used in biochemical and cell-based research.
  • Reported COX inhibitory and antiplatelet activity relevant to inflammation studies.
  • Demonstrates anti-inflammatory and anticancer effects in preclinical models.
  • Available as a high-purity solid suitable for analytical and biological assays.
  • Provided with datasheet, certificate of analysis, and safety data sheet for research compliance.
Catalog No. 50-002-89328