Learn More
TARGETMOL CHEMICALS INC 8-SHOGAOL 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T54305MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. [8]-Shogaol is a component of ginger and maintains anti-inflammatory activity as a cyclooxygenase-2 inhibitor. purity: 99%
Catalog No. 50-257-9766