Learn More
Cayman Chemical 8 Shydroxy9 RHexahydroca 100ug
SDP
Supplier:  Cayman Chemical 36248100 UG
Encompass_Preferred
An analytical reference standard that is structurally similar to known phytocannabinoids; a metabolite of hexahydrocannabinol; intended for research and forensic applications
Catalog No. 50-291-3060