Learn More
Cayman Chemical 8 Z 11 Z 14 ZHeptadecatrienal
SDP
Supplier:  Cayman Chemical 100051545 MG
Encompass_Preferred
A polyunsaturated fatty aldehyde; a degradation product of 2(R)-hydroperoxy linolenic acid; active against the plant pathogenic bacteria P. syringae; protects Arabidopsis plants against infection by the bacteria Pst DC3000 when injected into the leaves at 150 µM prior to bacterial inoculation
Catalog No. 50-289-0757