Learn More
MSC 80/20 Inc. Joining Plate, Aluminum, 12 Hole, 4 in Height, 0.1880 in Thickness
Supplier:  MSC 03779980
Encompass
This joining plate is constructed from aluminum with an anodized gray finish, designed for easy assembly using an external fastening method. It features 12 mounting holes, a thickness of 0.1880 inches, and an overall length and height of 4 inches.
  • External fastening method
  • Easily assembled
  • Tee flat plate design
  • Made of aluminum
  • Anodized gray finish
  • 12 mounting holes
  • 0.1880 in thickness
  • 4 in overall length and height
Catalog No. 50-005-29457