Learn More
Sino Biological Recombinant Rat GPT Protein (His Tag)
Supplier:  Sino Biological 80001R08B10
Encompass_Preferred
A DNA sequence encoding the rat Gpt1 (NP_112301.1) (Met 1-Ser 496) with a C-terminal polyhistidine tag was expressed.
Catalog No. 50-162-3514