Learn More
Sino Biological Recombinant Rat TIM-1/HAVCR1 Protein (hFc & His Tag)
Supplier:  Sino Biological 80003R16H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of rat HAVCR1 (NP_775172.1) (Ser 18-Val 238) was fused with a polyhistidine tag at the C-terminus and the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-3520