Learn More
Sino Biological Recombinant Rat c-MET/HGFR Protein (hFc Tag)
Supplier:  Sino Biological 80004R02H100
Encompass_Preferred
A DNA sequence encoding the rat MET (Q2IBC7) (Met1-Asn932) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3522