Learn More
Sino Biological Recombinant Rat PCSK9 Protein (His Tag)
Supplier:  Sino Biological 80005R08H20
Encompass_Preferred
A DNA sequence encoding the rat PCSK9 (NP_954862.2) (Met 1-Gln 691) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3525