Learn More
Sino Biological Recombinant Rat CD155/PVR Protein (His Tag)
Supplier:  Sino Biological 80007R08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of rat PVR (NP_058772.2) (Met 1-Gly 352) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3529