Learn More
Sino Biological Recombinant Rat RBP4 Protein (His Tag)
Supplier:  Sino Biological 80008R08H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain of rat RBP4 (NP_037294.1) (Met 1-Leu 201) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3532