Learn More
Sino Biological Recombinant Rat SOST/Sclerostin Protein (His Tag)
Supplier:  Sino Biological 80009R08H20
Encompass_Preferred
A DNA sequence encoding the rat SOST (NP_085073.1) (Met 1-Tyr 213) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3534