Learn More
Sino Biological Recombinant Rat CD89/FCAR Protein (His Tag)
Supplier:  Sino Biological 80014R08H100
Encompass_Preferred
A DNA sequence encoding the rat FCAR (NP_973721.1) extracellular domain (Met 1-Asn 228) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3543