Learn More
Sino Biological Recombinant Rat CD16a/FCGR3A/Fc gamma RIIIA Protein (His Tag), HPLC-verified
Supplier: Sino Biological 80017R08H100
A DNA sequence encoding the rat FCGR3A (XP_008767961.1) extracellular domain (Met 1-Pro 200) was expressed, fused with a polyhistidine tag at the C-terminus.