Learn More
Sino Biological Recombinant Rat CNTFR Protein (His Tag)
Supplier:  Sino Biological 80019R08B100
Encompass_Preferred
A DNA sequence encoding the rat CNTFR (Q08406-1) (Met 1-Pro 346) was fused with a C-terminal polyhistidine tag.
Catalog No. 50-162-3555