Learn More
Sino Biological Recombinant Rat GM-CSF/CSF2 Protein (His Tag)
Supplier:  Sino Biological 80020R07H50
Encompass_Preferred
A DNA sequence encoding the rat CSF2 (P48750) (Ala18-Lys144) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-3560