Learn More
Sino Biological Recombinant Rat GFR Alpha-1/GFRA1 Protein (hFc Tag)
Supplier:  Sino Biological 80021R02H100
Encompass_Preferred
A DNA sequence encoding the rat GFRA1 (Q62997) (Met1-Leu445) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3561