Learn More
Sino Biological Recombinant Rat ICAM-1/CD54 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 80022R02H100
Encompass_Preferred
A DNA sequence encoding the rat Icam1 (NP_037099.1) (Met1-Thr493) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3565