Learn More
Sino Biological Recombinant Rat B7-1/CD80 Protein (ECD, hFc Tag)
Supplier:  Sino Biological 80027R02H100
Encompass_Preferred
A DNA sequence encoding the rat Cd80 (AAB60503.1) (Met1-Gln248) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3578