Learn More
Sino Biological Recombinant Rat IL1R1 Protein (His & mFc Tag)
Supplier:  Sino Biological 80028R06H100
Encompass_Preferred
A DNA sequence encoding the rat IL1R1 (NP_037255.3) extracellular domain (Met 1-Lys 352) was was fused with the C-terminal His-tagged Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-162-3580