Learn More
Sino Biological Recombinant Rat IL1R1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 80028R08H100
Encompass_Preferred
A DNA sequence encoding the rat IL1R1 (NP_037255.3) extracellular domain (Met 1-Lys 352) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3582