Learn More
Sino Biological Recombinant Rat Growth Hormone Receptor Protein (His Tag)
Supplier:  Sino Biological 80029R08H100
Encompass_Preferred
A DNA sequence encoding the rat GHR (P16310-1) extracellular domain (Met 1-Arg 265) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3586