Learn More
Sino Biological Recombinant Rat gp130/IL6ST Protein (His Tag)
Supplier:  Sino Biological 80030R08H100
Encompass_Preferred
A DNA sequence encoding the rat IL6ST (NP_001008725.2) extracellular domain (Met 4-Glu 618) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3590