Learn More
Sino Biological Recombinant Rat ACE2 Protein (His Tag)
Supplier: Sino Biological 80031R08H20
A DNA sequence encoding the rat ACE2 (Q5EGZ1) extracellular domain (Met 1-Thr 740) was fused with a polyhistidine tag at the C-terminus.