Learn More
Sino Biological Recombinant Rat E-Selectin/CD62E Protein (hFc Tag)
Supplier:  Sino Biological 80033R02H100
Encompass_Preferred
A DNA sequence encoding the rat SELE (NP_620234.1) (Met1-Pro494) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3594