Learn More
Sino Biological Recombinant Rat E-Selectin/CD62E Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 80033R08H100
Encompass_Preferred
A DNA sequence encoding the rat SELE (P98105-1) extracellular domain (Met 1-Pro 494) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3596