Learn More
Sino Biological Recombinant Rat CXCL1 Protein
Supplier:  Sino Biological 80039RNCE100
Encompass_Preferred
A DNA sequence encoding the rat Cxcl1 (NP_110472.1) (Ala25-Lys96) was expressed.
Catalog No. 50-162-3602