Learn More
Sino Biological Recombinant Rat C-Reactive Protein Protein (His Tag)
Supplier: Sino Biological 80041R08H200
A DNA sequence encoding the rat CRP (NP_058792.1) extracellular domain (Met 1-Ser 230) was fused with a polyhistidine tag at the C-terminus.