Learn More
Sino Biological Recombinant Rat GRO Beta/CXCL2 Protein
Supplier:  Sino Biological 80042RNCE100
Encompass_Preferred
A DNA sequence encoding the rat Cxcl2 (NP_446099.1) (Ser32-Asn100) was expressed with two additional amino acids (Gly & Pro ) at the N-terminus.
Catalog No. 50-162-3609