Learn More
Sino Biological Recombinant Rat Fas/CD95/TNFRSF6 Protein (His Tag)
Supplier:  Sino Biological 80043R08H100
Encompass_Preferred
A DNA sequence encoding the rat FAS (NP_631933.2) extracellular domain (Met 1-Lys 170) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3614