Learn More
Sino Biological Recombinant Rat TNF-alpha Protein
Supplier:  Sino Biological 80045RNAE20
Encompass_Preferred
A DNA sequence encoding the mature form of rat TNFα (P16599) (Leu 80-Leu 235) was expressed and purified, with an initial Met at the N-terminus.
Catalog No. 50-162-3620