Learn More
Sino Biological Recombinant Rat L-selectin/CD62L Protein (His Tag)
Supplier:  Sino Biological 80046R08H100
Encompass_Preferred
A DNA sequence encoding the rat SELL (NP_062050.3) (Met1-Asn332) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3623