Learn More
Sino Biological Recombinant Rat NXPH1 Protein (His Tag)
Supplier:  Sino Biological 80047R08H100
Encompass_Preferred
A DNA sequence encoding the rat NXPH1 (Q63366-1) (Met 1-Gly 271) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3625