Learn More
Sino Biological Recombinant Rat IP-10/CXCL10 Protein (His Tag)
Supplier: Sino Biological 80057R08Y100
A DNA sequence encoding the rat CXCL10 (NP_620789.1) (Ile22-Pro98) was expressed with a polyhistidine tag at the C-terminus.