Learn More
Sino Biological Recombinant Rat Lipocalin-2/NGAL Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 80058R08B100
Encompass_Preferred
A DNA sequence encoding the rat LCN2 (P30152) (Met 1-Asn 198) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3637