Learn More
Sino Biological Recombinant Rat Serum Amyloid P Protein (His Tag)
Supplier:  Sino Biological 80061R08H100
Encompass_Preferred
A DNA sequence encoding the rat APCS (P23680) (Met 1-Ser 228) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3639