Learn More
Sino Biological Recombinant Rat Prolactin Receptor Protein (His Tag)
Supplier:  Sino Biological 80062R08H100
Encompass_Preferred
A DNA sequence encoding the rat PRLR (P05710-1) extracellular domain (Met 1-Asp 229) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3641