Learn More
Sino Biological Recombinant Rat TIE-2 Protein (hFc Tag)
Supplier:  Sino Biological 80063R02H200
Encompass_Preferred
A DNA sequence encoding the rat TEK (NP_001099207.1) extracellular domain (Met 4-Leu 743) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3644