Learn More
Sino Biological Recombinant Rat CXCL7/NAP-2 Protein (His Tag)
Supplier: Sino Biological 80066R08Y100
A DNA sequence encoding the rat Cxcl7 (NP_714943.1) (Lys38-Tyr111) was expressed with a polyhistidine tag at the C-terminus.