Learn More
Sino Biological Recombinant Rat PDGF-A Protein (His Tag)
Supplier:  Sino Biological 80068R07Y50
Encompass_Preferred
A DNA sequence encoding the rat Pdgfa (NP_036933.2) (Ser87-Arg196) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-3654