Learn More
Sino Biological Recombinant Rat IL-1RA/IL-1RN Protein (hFc Tag)
Supplier:  Sino Biological 80073R01H50
Encompass_Preferred
A DNA sequence encoding the rat IL1RN (P25086) (His27-Gln178) was expressed, fused with Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-3656