Learn More
Sino Biological Recombinant Rat CD25/IL-2R alpha Protein (His Tag), HPLC-verified
Supplier: Sino Biological 80074R08B200
A DNA sequence encoding the rat Il2ra (NP_037295.1) (Met1-Gln235) was expressed with a polyhistidine tag at the C-terminus.