Learn More
Sino Biological Recombinant Rat IL-2 Protein
Supplier: Sino Biological 80075RNAE100
A DNA sequence encoding the rat IL2 (P17108) (Ala 21-Gln 155) was expressed and purified, with an initial Met at the N-terminus.