Learn More
Sino Biological Recombinant Rat IL-6 Protein
Supplier:  Sino Biological 80076RNAE100
Encompass_Preferred
A DNA sequence encoding the rat IL6 (NP_036721.1)(Phe25-Thr211) was expressed and purified with an initial Met.
Catalog No. 50-162-3662