Learn More
Sino Biological Recombinant Rat IL9R Protein (His Tag)
Supplier: Sino Biological 80077R08H50
A DNA sequence encoding the rat IL9R / Interleukin 9 receptor (EDM04022.1) extracellular domain (Met 1-Ala 270) was expressed, fused with a polyhistidine tag at the C-terminus.