Learn More
Sino Biological Recombinant Rat Her2/ERBB2 Protein (hFc Tag)
Supplier: Sino Biological 80079R02H100
A DNA sequence encoding the rat ErbB2 (P06494) extracellular domain (Met1-Thr654) was fused with the Fc region of human IgG1 at the C-terminus.